Learn More
Aviva Systems Biology Corp OR1I1 Antibody - C-terminal region (ARP59698_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59698_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR1I1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR1I1. Alternate names: OR19-20, OR1I1P, OR1I1QApplicationWB. Species reactivity: Human
Catalog No. 50-286-783