Learn More
Aviva Systems Biology Corp OR1L8 Antibody - C-terminal region (ARP59701_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59701_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR1L8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR1L8. Alternate names: OR9-24ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-785