Learn More
Aviva Systems Biology Corp OR1S1 Antibody - C-terminal region (ARP59708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR1S1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR1S1. Alternate names: OR11-232, OST034ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-786