Learn More
Aviva Systems Biology Corp OR2B2 Antibody - C-terminal region (ARP59715_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59715_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2B2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2B2. Alternate names: OR2B2Q, OR2B9, OR6-1, dJ193B12.4, hs6M1-10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-789