Learn More
Aviva Systems Biology Corp OR2C1 Antibody - C-terminal region (ARP59716_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59716_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2C1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2C1. Alternate names: MGC163200, MGC95444, OLFmf3, OR2C2PApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-790