Learn More
Aviva Systems Biology Corp OR2C3 Antibody - C-terminal region (ARP59718_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59718_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2C3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2C3. Alternate names: OR2C4, OR2C5P, OST742ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-286-791