Learn More
Aviva Systems Biology Corp OR2H1 Antibody - C-terminal region (ARP59729_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59729_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2H1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2H1. Alternate names: 6M1-16, HS6M1-16, OLFR42A-9004-14, OR2H6, OR2H8, OR6-2, dJ994E9.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-793