Learn More
Aviva Systems Biology Corp OR2H2 Antibody - C-terminal region (ARP59731_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59731_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2H2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2H2. Alternate names: FAT11, MGC126732, MGC138381, OLFR2, OLFR42B, OR2H3, dJ271M21.2, hs6M1-12ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-286-795