Learn More
Aviva Systems Biology Corp OR2J2 Antibody - C-terminal region (ARP59732_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59732_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2J2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2J2. Alternate names: MGC119134, MGC119135, MGC119136, MGC119137, OR6-19, OR6-8, OR6.3.8, ORL684, dJ80I19.4, hs6M1-6ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit
Catalog No. 50-286-796