Learn More
Aviva Systems Biology Corp OR2K2 Antibody - C-terminal region (ARP59734_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59734_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2K2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2K2. Alternate names: HSHTPCRH06, HTPCRH06, MGC133152, OR2AN1P, OR2AR1PApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-798