Learn More
Aviva Systems Biology Corp OR2M2 Antibody - C-terminal region (ARP59740_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59740_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2M2. It was validated on Western Blot. . Alternate names: OR2M2Q, OST423ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-286-802