Learn More
Aviva Systems Biology Corp OR2W1 Antibody - C-terminal region (ARP59797_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59797_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2W1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OR2W1. Alternate names: MGC119162, MGC119163, MGC119165, hs6M1-15ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-829