Learn More
Aviva Systems Biology Corp OR2W1 Antibody - C-terminal region (ARP59798_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59798_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2W1. It was validated on Western Blot. . Alternate names: MGC119162, MGC119163, MGC119165, hs6M1-15ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-286-830