Learn More
Aviva Systems Biology Corp OR2W5 Antibody - middle region (ARP44489_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44489_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OR2W5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OR2W5. Alternate names: OR2W5P, OST722ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53893