Learn More
Instech Polyurethane feeding tubes, 13ga x 88mm, 10 sterile pouches of 5 (50 tubes)
SDP
Catalog No. 5000510042
Encompass
The Instech FTPU-13-88-50 is a sterile 13 gauge polyurethane feeding tube designed for oral gavage in rats It features a soft polyurethane tube with a straight rounded tip engineered to minimize esophageal trauma and reduce the risk of inadvertent tracheal placement The tube includes a clear medical grade copolyester hub and offers a larger internal diameter to support delivery of viscous compounds FTPU 13 88 50 is supplied in 10 sterile pouches of 50 sterile pouches of 5 (250 tubes) br br Features ul li Gauge 13ga /li li Length 88 mm /li li Tube Material Polyurethane /li li Tip Material Rounded trauma reducing /li li Connection Clear medical grade copolyester hub /li li Sterility EtO sterilized /li li Package Configuration 50 sterile pouches of 5 (250 tubes) /li li Use Case Rat oral gavage for dosing and compound delivery /li /ul
Catalog No. 50-005-10042 Supplier Instech Supplier No. FTPU1388
The Instech FTPU-13-88-50 is a sterile 13 gauge polyurethane feeding tube designed for oral gavage in rats It features a soft polyurethane tube with a straight rounded tip engineered to minimize esophageal trauma and reduce the risk of inadvertent tracheal placement The tube includes a clear medical grade copolyester hub and offers a larger internal diameter to support delivery of viscous compounds FTPU 13 88 50 is supplied in 10 sterile pouches of 50 sterile pouches of 5 (250 tubes) br br Features ul li Gauge 13ga /li li Length 88 mm /li li Tube Material Polyurethane /li li Tip Material Rounded trauma reducing /li li Connection Clear medical grade copolyester hub /li li Sterility EtO sterilized /li li Package Configuration 50 sterile pouches of 5 (250 tubes) /li li Use Case Rat oral gavage for dosing and compound delivery /li /ul