Learn More
Instech Plastic feeding tubes, 22ga (black) x 38mm, 10 sterile pouches of 5 (50 tubes)
SDP
Catalog No. 5000510044
Encompass
The Instech FTP-22-38-50 is a sterile 22 gauge polypropylene feeding tube designed for oral gavage in small rodents It features a flexible translucent polypropylene tube with a black female luer hub and a soft elastomer tip engineered to minimize esophageal trauma and reduce the risk of inadvertent tracheal placement FTP 22 38 50 is supplied in 50 sterile pouches of 5 (250 tubes) br br Features ul li Gauge 22ga (black hub) /li li Length 38 mm /li li Tube Material Medical-grade polypropylene /li li Tip Material Soft elastomer trauma reducing /li li Connection Female luer /li li Sterility EtO sterilized /li li Package Configuration 50 sterile pouches of 5 (250 tubes) /li li Use Case Rodent oral gavage for dosing and compound delivery /li /ul
Catalog No. 50-005-10044 Supplier Instech Supplier No. FTP2238
The Instech FTP-22-38-50 is a sterile 22 gauge polypropylene feeding tube designed for oral gavage in small rodents It features a flexible translucent polypropylene tube with a black female luer hub and a soft elastomer tip engineered to minimize esophageal trauma and reduce the risk of inadvertent tracheal placement FTP 22 38 50 is supplied in 50 sterile pouches of 5 (250 tubes) br br Features ul li Gauge 22ga (black hub) /li li Length 38 mm /li li Tube Material Medical-grade polypropylene /li li Tip Material Soft elastomer trauma reducing /li li Connection Female luer /li li Sterility EtO sterilized /li li Package Configuration 50 sterile pouches of 5 (250 tubes) /li li Use Case Rodent oral gavage for dosing and compound delivery /li /ul