Learn More
GLOMED INC ORAL GAVAGE NEEDLE 16G X76MM
Catalog No. NC2203923
Encompass_Preferred
Reusable Oral Gavage Needle (Pack of 1) - 16G x 76mm (3mm Tip Diameter) / Curved (Product is through Pet Surgical)
This item is not returnable. View return policy
Catalog No. NC2203923 Supplier GLOMED INC Supplier No. AFN1676C
Reusable Oral Gavage Needle (Pack of 1) - 16G x 76mm (3mm Tip Diameter) / Curved (Product is through Pet Surgical)