Learn More
GLOMED INC ORAL GAVAGE NEEDLE,20G,20/BX
Catalog No. NC1906313
Encompass_Preferred
Disposable Oral Gavage Needle (Box of 20) (Sterile)20G x 38mm (2mm Tip) Curved
This item is not returnable. View return policy
Catalog No. NC1906313 Supplier GLOMED INC Supplier No. DAFN2038C
 Restricted Item
Add to Cart
Add to Cart
Disposable Oral Gavage Needle (Box of 20) (Sterile)20G x 38mm (2mm Tip) Curved