Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sol-M™ Oral Syringe

Catalog No. 22024332
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
Color:
Amber
Clear
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
22-024-332 3 mL Clear
22-024-329 1 mL Amber
22-024-330 1 mL Clear
22-024-331 3 mL Amber
22-024-333 5 mL Amber
22-024-334 5 mL Clear
22-024-335 10 mL Amber
22-024-336 10 mL Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-024-332 Supplier Sol-M™ Supplier No. 21003OC
Add to Cart
Add to Cart
This item is not returnable. View return policy

Oral syringe specifically for use with oral medications and featuring an o-ring design.

  • Large variety of sizes and colors
  • Clear graduation markings for accurate medication delivery
  • Not made with natural rubber latex
  • Special tip design will not accept Luer connectors, preventing medication errors and the accidental intravenous injections of oral liquids
  • Optional amber barrel designed for light-sensitive drugs, in compliance with United States Pharmacopeia (USP) requirements

Specifications

Autoclavable Not Autoclavable
Material Polypropylene
Color Clear
Needle Gauge No Needle Included
Includes Tip Cap
Latex Free Yes
Disposable Single-use
Packaging Blister
Volume (Metric) 3 mL
For Use With (Equipment) Oral Syringe
Connection Type Catheter
Graduated Yes
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.