Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oralith Brilliant Pink R 95.0+%, TCI America™
SDP

Catalog No. O012925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O012925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O012925G Supplier TCI America Supplier No. O012925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2379-74-0
Molecular Formula C18H10Cl2O2S2
Molecular Weight (g/mol) 393.30
MDL Number MFCD00059165
InChI Key NDDLLTAIKYHPOD-ZCXUNETKSA-N
Synonym Threne Brilliant Pink R, Vat Red 1
PubChem CID 3000709
IUPAC Name 6-chloro-2-[(2Z)-6-chloro-4-methyl-3-oxo-2,3-dihydro-1-benzothiophen-2-ylidene]-4-methyl-2,3-dihydro-1-benzothiophen-3-one
SMILES CC1=CC(Cl)=CC2=C1C(=O)\C(S2)=C1\SC2=CC(Cl)=CC(C)=C2C1=O

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 393.30
Percent Purity ≥95.0% (E)
Chemical Name or Material Oralith Brilliant Pink R
EINECSNumber (5)-2207
RTECSNumber DL9825500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.