Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 9178NFO Orange Aramid Fiber Gloves, Nitrile Foam Coating

Catalog No. 19812639 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-639 2X-Large
19-812-634 X-Small
19-812-635 Small
19-812-636 Medium
19-812-637 Large
19-812-638 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-812-639 Supplier MCR Safety Supplier No. 9178NFOXXL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Orange gloves made from aramid fiber feature orange nitrile foam coating on palm and fingertips for secure wet or dry grip. High-visibility color enhances safety awareness.

  • High-visibility orange Kevlar™ aramid fiber engineered yarn
  • Glove color enhances safety awareness
  • Lightweight seamless shell provides dexterity and comfort
  • Orange nitrile foam coating allows for tacky grip and sense of touch
  • Touch screen friendly

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Orange
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Manufacturing
Glove Cuff Style Knit
Hem Color Multi-colored
Model 9178NFOXXL
Series Cut Pro™ 9178NFO
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.