Learn More
Checkers Orange Aviation Wheel Chocks for Small Aircraft, Roped Pair
GREENER_CHOICE
Catalog No. 19088421 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Orange
Catalog No. Color
19-088-421 Orange
1 options
Catalog No. 19-088-421 Supplier Checkers Supplier No. UC200S
Add to Cart
Add to Cart
Fits the curvature of the tire to absorb wheel-squat during loading and fueling operations. Features molded-in soft base for increased traction. Replaceable polyester rope lanyard connects two chocks together. Custom rope lengths available. Sold in pairs.
  • For small aircraft
  • Impact-absorbing, lightweight urethane
  • Weather and abrasion resistant
  • High load-bearing capacity
  • Resistant to aircraft fuels, oils, and lubricants
  • Eliminates Foreign Object Damage (FOD)
  • Will not rot, crack or splinter like wood
  • Sold roped together as pairs
  • Molded-in high-visibility yellow color
Specifications
Product Type Wheel Chock
Material Urethane
Color Orange