Learn More
Perkin Elmer US LLC Orange, Capillary split/splitless deactivated glass liners with deactivated wool. Quartz wool is used to fully vaporize the sample.
Catalog No. 502104649
Encompass_Preferred
Ideal for general purpose applications Quartz Wool is used to fully vaporize the sample Color coded ORANGE on the liner with PKI name
Catalog No. 50-210-4649 Supplier Perkin Elmer US LLC Supplier No. N9306236
Ideal for general purpose applications Quartz Wool is used to fully vaporize the sample Color coded ORANGE on the liner with PKI name