Learn More
Andwin Scientific Orange Cryo-Babies 1.28" x 0.50"
SDP
Catalog No. NC9549070
Encompass_Preferred
Cryogenic labels formatted for laser printers. The 1.28" x 0.50" Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of orange.
Catalog No. NC9549070 Supplier Andwin Scientific Supplier No. LCRY1700O
Cryogenic labels formatted for laser printers. The 1.28" x 0.50" Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of orange.