Learn More
Andwin Scientific Orange Cryo labels 1.69 x 0.75"
SDP
Supplier:  Andwin Scientific LCRY1200O
Encompass_Preferred
Cryogenic labels formatted for laser printers. The Cryo-Tags labels measure 1.69 x 0.75". This product features compatibility with laser printers as the labels are heat-resistant and have a stay-flat design. The labels can withstand temperatures from -196C to 100C and can edure boiling water baths, oven temperatures, and autoclaving and freezer temperatures. The Cryo-Tags can also combat most organic solvents and caustic agents. Compatible with most surfaces, like plastics, glass, and metals without cracking, peeling, or degrading. This is sold in the color of orange.
Catalog No. NC9549072