Learn More
BERTECH KELEX Industrial Grade Finger Cots, Ultra Tough, 14 Mil Thick, Large (300 Pcs./Bag)
SDP
Catalog No. NC1570679
Encompass_Preferred
Industrial Grade Finger Cots are made of heavy 14 mil natural rubber latex material. These finger cots are tough, durable, and re-usable. They are primarily used where sharp objects are involved. These heavy duty finger cots are unrolled, lightly powdered, and textured for a better grip. They are packaged as 300 pieces per bag.Applications: Automotive, Banking, Electronic Assembly, Industrial and retail facilities.
This item is not returnable. View return policy
Catalog No. NC1570679 Supplier BERTECH KELEX Supplier No. FCOL
Industrial Grade Finger Cots are made of heavy 14 mil natural rubber latex material. These finger cots are tough, durable, and re-usable. They are primarily used where sharp objects are involved. These heavy duty finger cots are unrolled, lightly powdered, and textured for a better grip. They are packaged as 300 pieces per bag.Applications: Automotive, Banking, Electronic Assembly, Industrial and retail facilities.