Learn More
Bioworld Orange-G Blue Loading Dye (6X), 10 mL
Catalog No. 50196791
Encompass_Preferred
Orange-G Loading Dye, Blue, 6X Gel loading dye used in molecular biology protocols. Technical Information:6X concentrationOrange-G (0.25%)Bromophenol blue (0.25%)Xylene cyanol FF (0.25%)Ficol in water (30%)EDTA (30mM)
Catalog No. 50-196-791 Supplier Bioworld Supplier No. 105700251
Orange-G Loading Dye, Blue, 6X Gel loading dye used in molecular biology protocols. Technical Information:6X concentrationOrange-G (0.25%)Bromophenol blue (0.25%)Xylene cyanol FF (0.25%)Ficol in water (30%)EDTA (30mM)