Learn More
Electron Microscopy Sciences Orange G, Certified, C.I. DcO-13 25 GR
SDP
Catalog No. 5027564
Encompass_Preferred
(Acid Orange 10, Wood Orange 2G)
C₁₆H₁0N₂O7S₂Na₂ F.W. 452.38
Solubility: 8.0% Water; 0.22% Alcohol; 1.7% Cellosolve; 3.0% Glycol; 0.0% Xylene
Orange G or Kornhauser's Quad stain is for most elementary structures of tissues. Stain Tech., 18, 985 (1943) Mallory's tissue stain for connective tissue. Mallory, J. Exper. Med., 5,15 (1900). Used with safranin and tannic acid for showing shoot apex in plant tissue. Stain Tech., 18, 105 (1943).
Catalog No. 50-275-64 Supplier Electron Microscopy Sciences Supplier No. 19070
(Acid Orange 10, Wood Orange 2G)
C₁₆H₁0N₂O7S₂Na₂ F.W. 452.38
Solubility: 8.0% Water; 0.22% Alcohol; 1.7% Cellosolve; 3.0% Glycol; 0.0% Xylene
Orange G or Kornhauser's Quad stain is for most elementary structures of tissues. Stain Tech., 18, 985 (1943) Mallory's tissue stain for connective tissue. Mallory, J. Exper. Med., 5,15 (1900). Used with safranin and tannic acid for showing shoot apex in plant tissue. Stain Tech., 18, 105 (1943).