Learn More
AG Scientific Inc Orange G Loading Dye 25 ML
Catalog No. 502268342
Encompass_Preferred
Orange G (0.25%), Glycerol (30%) in sterile deionized water. 6X Concentrated loading dye for molecular biology applications. It is a pre-mixed solution used to prepare DNA-markers and samples.Used for tracking DNA migration during electrophoresis on polyacrylamide gels or agarose.
Catalog No. 50-226-8342 Supplier AG Scientific Inc Supplier No. O257325ML
Orange G (0.25%), Glycerol (30%) in sterile deionized water. 6X Concentrated loading dye for molecular biology applications. It is a pre-mixed solution used to prepare DNA-markers and samples.Used for tracking DNA migration during electrophoresis on polyacrylamide gels or agarose.