Learn More
Bioworld Orange-G Loading Dye with Glycerol (6X), 25 mL
Catalog No. 50196789
Encompass_Preferred
Orange-G Loading Dye, Glycerol, 6X Gel loading dye used in molecular biology protocols. Technical Information:6X concentrationOrange-G (0.25%)Glycerol in water (30%)
Catalog No. 50-196-789 Supplier Bioworld Supplier No. 105700241
Orange-G Loading Dye, Glycerol, 6X Gel loading dye used in molecular biology protocols. Technical Information:6X concentrationOrange-G (0.25%)Glycerol in water (30%)