Learn More
Bioworld Orange-G Loading Dye with Glycerol (6X), 50 mL
Catalog No. 50196790
Encompass_Preferred
Orange-G Loading Dye, Glycerol, 6X Gel loading dye used in molecular biology protocols. Technical Information:6X concentrationOrange-G (0.25%)Glycerol in water (30%)
Catalog No. 50-196-790 Supplier Bioworld Supplier No. 105700242
Orange-G Loading Dye, Glycerol, 6X Gel loading dye used in molecular biology protocols. Technical Information:6X concentrationOrange-G (0.25%)Glycerol in water (30%)