Learn More
Bioworld Orange G sodium salt, 100 g
Supplier:  Bioworld 217700282
Encompass_Preferred
- High Purity Grade Dye Content(%)- >80.0 Lambda Max(nm)- 472-478 Loss on Drying(%)- Report Solubility (0.002%, Water)(P/F)- Pass
Catalog No. 50-199-231