Learn More
Electron Microscopy Sciences Orange G Solution 500 ML
SDP
Catalog No. 5031823
Encompass_Preferred
Used in Dane's Method for Prekeratin, Keratin, and Mucin. Keratin stain method for the staining of keratin, prekeratin, acid mucopolysaccharides and nuclei. See Dane, E.T., and Herman, D.L.: Stain Technology, 38:97, 1963., or AFIP Manual of Histological Staining Methods, 3rd Ed., Ed L. Luna: New York: McGraw Hill Publications, c 1968, p.83.
Catalog No. 50-318-23 Supplier Electron Microscopy Sciences Supplier No. 2640103
Used in Dane's Method for Prekeratin, Keratin, and Mucin. Keratin stain method for the staining of keratin, prekeratin, acid mucopolysaccharides and nuclei. See Dane, E.T., and Herman, D.L.: Stain Technology, 38:97, 1963., or AFIP Manual of Histological Staining Methods, 3rd Ed., Ed L. Luna: New York: McGraw Hill Publications, c 1968, p.83.