Learn More
Medchemexpress LLC Orange I | 523-44-4 | 86.3% | 250 MG
SDP
Catalog No. 5000414456
Encompass_Preferred
Orange I (α-Naphthol Orange) is an anionic azo dye primarily used in research. It is known for its ability to form a complex with Methylene blue, resulting in fluorescence quenching and alterations to the absorption spectra of both compounds. The equilibrium constant for this complex formation is 79900 mol-1·dm3.
  • Anionic azo dye
  • Forms a complex with Methylene blue
  • Causes fluorescence quenching
  • Changes absorption spectrum of itself and Methylene blue
  • Equilibrium constant of the complex is 79900 mol-1·dm3
  • For research use only
  • Can be dissolved in deionized water or suitable buffers
  • Used in spectroscopy for absorption spectrum analysis
  • Useful for visual and colorimetric detection
  • Applicable for adsorption capacity determination
Catalog No. 50-004-14456 Supplier Medchemexpress LLC Supplier No. 250MGHYD0929
Add to Cart
Add to Cart
Orange I (α-Naphthol Orange) is an anionic azo dye primarily used in research. It is known for its ability to form a complex with Methylene blue, resulting in fluorescence quenching and alterations to the absorption spectra of both compounds. The equilibrium constant for this complex formation is 79900 mol-1·dm3.
  • Anionic azo dye
  • Forms a complex with Methylene blue
  • Causes fluorescence quenching
  • Changes absorption spectrum of itself and Methylene blue
  • Equilibrium constant of the complex is 79900 mol-1·dm3
  • For research use only
  • Can be dissolved in deionized water or suitable buffers
  • Used in spectroscopy for absorption spectrum analysis
  • Useful for visual and colorimetric detection
  • Applicable for adsorption capacity determination