Learn More
Electron Microscopy Sciences Orange II 25 GR
SDP
Catalog No. 5027565
Encompass_Preferred
(Acid Orange 7; Acid Orange II, Y or A; Gold Orange; Mandarin G; Orange A, P or R; Orange Extra; Tropaoelin OOO No.2)
C₁₆H₁₁N₂O₄Sna F.W. 350.33
Solubility: 3.0% Water; 0.15% Alcohol; 2.5% Cellosolve; 8.0% Glycol; 0.0% Xylene Dye Content: approx. 75%
Orange II is a general tissue stain with Eosin and Azur-C. French, Stain Tech., 1, 79 (1926). Counterstain for Heidenhain's Hematoxylin in animal histology. A substitute for Orange G if a stronger yellow is desired for contrast. J. Lab. Med., 28, 995 (1943)
Catalog No. 50-275-65 Supplier Electron Microscopy Sciences Supplier No. 19060
(Acid Orange 7; Acid Orange II, Y or A; Gold Orange; Mandarin G; Orange A, P or R; Orange Extra; Tropaoelin OOO No.2)
C₁₆H₁₁N₂O₄Sna F.W. 350.33
Solubility: 3.0% Water; 0.15% Alcohol; 2.5% Cellosolve; 8.0% Glycol; 0.0% Xylene Dye Content: approx. 75%
Orange II is a general tissue stain with Eosin and Azur-C. French, Stain Tech., 1, 79 (1926). Counterstain for Heidenhain's Hematoxylin in animal histology. A substitute for Orange G if a stronger yellow is desired for contrast. J. Lab. Med., 28, 995 (1943)