Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Orange light-emitting PPV copolymer
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Features:Soluble orange light-emitting PPV copolymer.Excellent reproducibility.Broad emission spectrum.High efficiency.Very low operation voltage.Very long DC and AC lifetime.High temperature stability (>100°C).Typical performance measured in standard device stack (ITO/PEDOT:PSS 20 nm/PDO-123 80 nm/Ba 6 nm/Ag 150 nm):Max. efficiency: 5.0 Cd/A.V_on: 2.0 V.V at 100 cd/m2: 2.6 V.V at 1000 cd/m2: 3.3 V.Max. power eff.: 5.5 lm/W.External quantum eff.: 3.5%.CIE1931 at 100 cd/m2: (0.60, 0.40).DC lifetime at 100 cd/m2 at RT: >>25000.Product of EMD Performance Materials Corp., an Affiliate of Merck KGaA, Darmstadt, Germany
Specifications
Specifications
| CAS | 1351337-33-1 |
| Quantity | 1 g |
| Synonym | Livilux(TM) PDO-123 |
| Molecular Weight (g/mol) | 1750000 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?