Learn More
Novus Biologicals™ ORANGE Peroxidase Substrate Kit

Specifications
Specifications
| Product Type | Substrate Kit |
| Quantity | 1 Kit |
| For Use With (Application) | Immunohistochemistry (IHC) |
| Sample Type | Biological Sample |
| Target Specificity | ORANGE Peroxidase |
| Content And Storage | Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.