Learn More
Medchemexpress LLC Orantinib | 252916-29-3 | 98.5% | 310.35 | C18H18N2O3 | 5 MG
SDP
Catalog No. 5000242543
Encompass_Preferred
Orantinib (SU6668; TSU-68) is a small-molecule, multi-targeted receptor tyrosine kinase inhibitor used in preclinical research. It inhibits VEGFR (Flt-1), PDGFRβ, and FGFR1 with reported Kis of 2.1 μM, 8 nM, and 1.2 μM, respectively. The compound has molecular formula C18H18N2O3, molecular weight 310.35, CAS 252916-29-3, and is provided as a purified research reagent.
  • Inhibits VEGFR (Flt-1), PDGFRβ, and FGFR1 with documented Ki values.
  • Exhibits antiangiogenic and antitumor activity in preclinical studies.
  • Supplied as a purified research-grade small molecule with reported purity around 98.5%.
  • Available in multiple sizes suitable for assay development and in vivo dosing studies.
  • Suitable for target validation, pathway studies, and pharmacology experiments.
Catalog No. 50-002-42543 Supplier Medchemexpress LLC Supplier No. HY105175MG
Orantinib (SU6668; TSU-68) is a small-molecule, multi-targeted receptor tyrosine kinase inhibitor used in preclinical research. It inhibits VEGFR (Flt-1), PDGFRβ, and FGFR1 with reported Kis of 2.1 μM, 8 nM, and 1.2 μM, respectively. The compound has molecular formula C18H18N2O3, molecular weight 310.35, CAS 252916-29-3, and is provided as a purified research reagent.
  • Inhibits VEGFR (Flt-1), PDGFRβ, and FGFR1 with documented Ki values.
  • Exhibits antiangiogenic and antitumor activity in preclinical studies.
  • Supplied as a purified research-grade small molecule with reported purity around 98.5%.
  • Available in multiple sizes suitable for assay development and in vivo dosing studies.
  • Suitable for target validation, pathway studies, and pharmacology experiments.