Learn More
Selleck Chemical LLC Orantinib (SU6668) S1470-5mg
SDP
Catalog No. 503108538
Encompass_Preferred
Orantinib (TSU-68 SU6668) has greatest potency against PDGFR autophosphorylation with Ki of 8 nM in a cell-free assay but also strongly inhibits Flk-1 and FGFR1 trans-phosphorylation little activity against IGF-1R Met Src Lck Zap70 Abl and CDK2 does not inhibit EGFR Phase 3
Catalog No. 50-310-8538 Supplier Selleck Chemical LLC Supplier No. S14705MG
Orantinib (TSU-68 SU6668) has greatest potency against PDGFR autophosphorylation with Ki of 8 nM in a cell-free assay but also strongly inhibits Flk-1 and FGFR1 trans-phosphorylation little activity against IGF-1R Met Src Lck Zap70 Abl and CDK2 does not inhibit EGFR Phase 3