Learn More
Medchemexpress LLC HY-10517 50mg Medchemexpress, Orantinib CAS:252916-29-3 Purity:>98%
SDP
Catalog No. 501968253
Encompass_Preferred
Medchemexpress, HY-10517 50mg Orantinib CAS:252916-29-3 Orantinib (SU6668; TSU-68) is a multi-targeted receptor tyrosine kinase inhibitor with Kis of 2.1 μM, 8 nM and 1.2 μM for Flt-1, PDGFRβ and FGFR1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8253 Supplier Medchemexpress LLC Supplier No. HY1051750MG
Medchemexpress, HY-10517 50mg Orantinib CAS:252916-29-3 Orantinib (SU6668; TSU-68) is a multi-targeted receptor tyrosine kinase inhibitor with Kis of 2.1 μM, 8 nM and 1.2 μM for Flt-1, PDGFRβ and FGFR1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.