Learn More
OraSure Technologies OraQuick™ ADVANCE™ Kit Controls
SDP
Catalog No. 23222102 Shop All OraSure Technologies Products
Change view
Click to view available options
CE Marker:
Yes
Catalog No. CE Marker
23-222-102 Yes
1 options
This item is not returnable. View return policy
Catalog No. 23-222-102 Supplier OraSure Technologies Supplier No. 10010077
Add to Cart
Add to Cart
This item is not returnable. View return policy
The OraQuick™ ADVANCE™ Rapid HIV-1/2 Antibody Test Kit Controls are quality control reagents for use only with the OraQuick™ ADVANCE™ Rapid HIV-1/2 Antiboby Test.
  • Each Kit Control Box contains a package insert and three vials (one HIV-1 positive control, one HIV-2 positive control and one negative control).
  • Store the controls at 2 to 8 degrees C (35 to 46 degrees F).
  • One black-capped vial containing 0.2 mL of photochemically inactivated human plasma positive for antibodies to HIV-1, diluted in a defibrinated pool of normal human plasma.
  • One red-capped vial containing 0.2 mL of photochemically inactivated human plasma positive for antibodies to HIV-2, diluted in a defibrinated pool of normal human plasma.
  • One white-capped vial containing 0.2 mL of photochemically inactivated human plasma negative for antibodies to HIV-1 and HIV-2.
  • Negative for Hepatitis B surface antigen and Hepatitis C antibody.
Run the Kit Controls under the following circumstances:
  • Each new operator prior to performing testing on patient specimens
  • When opening a new test kit lot
  • Whenever a new shipment of test kits is received
  • If the temperature of the test kit storage area falls outside of 2-27°C (36-80°F)
  • If the temperature of the testing area falls outside of 15-37°C (59-99°F)
  • At periodic intervals as dictated by the user facility.
Specifications
Content And Storage 2°C to 8°C
CE Marker Yes
Disposable Single-use
Includes Each Kit Control Box contains a Pk. insert and three vials (one HIV-1 positive control, one HIV-2 positive control and one negative control).