Learn More
OraSure Technologies OraQuick™ HCV Antibody Test
SDP
Catalog No. 23222104 Shop All OraSure Technologies Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-222-104 100 Tests
23-222-105 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-222-104 Supplier OraSure Technologies Supplier No. 10010180
Add to Cart
Add to Cart
This item is not returnable. View return policy
HCV Testing anytime, anywhere made easy. The OraQuick™ HCV test is the only point-of-care Hepatitis C test on the market today, detecting HCV antibodies in fingerstick and venipuncture whole blood. Yielding accurate results in as little as 20 minutes.
  • The OraQuick™ HCV test is FDA approved for detecting HCV antibodies in fingerstick and venipuncture whole blood.
  • Our simple platform enables healthcare providers to deliver an accurate result in 20 minutes.
  • The OraQuick™ HCV Rapid Antibody Test is comprised of both a single-use test device and vial containing a pre-measured amount of a buffered developer solution.
  • The test consists of a sealed pouch with two separate compartments for each component.
  • The developer solution facilitates the capillary flow of the specimen into the device and onto the assay strip.
  • 25 Test Kit includes: 25 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
  • 100 Test Kit includes: 100 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
Specifications
Content And Storage 2°C to 30°C
Description Contains synthetic peptides and recombinant antigen from the core, NS3 and NS4 regions of the HCV genome.
CE Marker Yes
Detectable Analytes HCV
Disposable Single-use
Quantity 100 Tests