Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OraSure Technologies OraQuick™ HCV Control Solution Kit - Hepatitis C Antibody Test (Positive Level / Negative Level)
SDP

Catalog No. 23222106 Shop All OraSure Technologies Products
Change view
Click to view available options
Content And Storage:
2°C to 8°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
23-222-106 2°C to 8°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-222-106 Supplier OraSure Technologies Supplier No. 10010182
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The OraQuick™ HCV Rapid Antibody Test Kit Controls are quality control reagents for use only with the OraQuick™ HCV Rapid Antibody Test.

Run the Kit Controls under the following circumstances:

  • Each new operator prior to performing testing on patient specimens.
  • When opening a new test kit lot.
  • Whenever a new shipment of test kits is received.
  • If the temperature of the test kit storage area falls outside of 2-30°C (36-86°F).
  • If the temperature of the testing area falls outside of 15-37°C (59-99°F).
  • At periodic intervals as dictated by the user facility.

Package contains: HCV Positive Control (1 vial, purple cap, 0.2 mL), HCV Negative Control (1 vial, white cap, 0.2 mL), and Package Insert

Specifications

Content And Storage 2°C to 8°C
Includes HCV Positive Control (1 vial, purple cap, 0.2 mL), HCV Negative Control (1 vial, white cap, 0.2 mL), and Pk. Insert
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.