Learn More
OraSure Technologies OraQuick™ HCV Visual Reference Panel
SDP
Catalog No. 23222107 Shop All OraSure Technologies Products
Change view
Click to view available options
Quantity:
1 Set/Ea.
Catalog No. Quantity
23-222-107 1 Set/Ea.
1 options
This item is not returnable. View return policy
Catalog No. 23-222-107 Supplier OraSure Technologies Supplier No. 10010343
Add to Cart
Add to Cart
This item is not returnable. View return policy
The OraQuick™ HCV Visual Reference Panel is intended to assist new operators in becoming proficient at reading specimens with antibody levels near the limit of detection of the device.
OraQuick™ HCV Visual Reference Panel The Foil pouch contains a package insert and three devices (one HCV Limit of Detection Device, one HCV Low Reactive Device and one HCV Non-Reactive Device) as described below:
  • HCV Limit of Detection Device One OraQuick™ HCV Rapid Antibody Test device that has been manufactured at a predetermined reactivity level to produce a reactive test result.
  • HCV Low Reactive Device One OraQuick™ HCV Rapid Antibody Test device that has been manufactured at a predetermined reactivity level to produce a reactive test result.
  • HCV Non-Reactive Device One OraQuick™ HCV Rapid Antibody Test device that has been manufactured to produce a non-reactive test result.
Specifications
Content And Storage 15°C to 30°C
Disposable Single-use
Quantity 1 Set/Ea.