Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OraSure Technologies OraQuick™ HIV Self-Test
SDP

Catalog No. 22899026 Shop All OraSure Technologies Products
Change view
Click to view available options
Disposable:
Single-use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
22-899-026 Single-use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-899-026 Supplier OraSure Technologies Supplier No. 10010696
Add to Cart
Add to Cart

The OraQuick™ HIV Self-Test is the first and only FDA-approved oral fluid rapid, over-the-counter (OTC) test capable of detecting antibodies to HIV-1 and HIV-2.

OraQuick™ HIV Self-Test is the same trusted test used by healthcare providers in the comfort of your own space. The test is approved for use in individuals 14 years of age and older.

Time to result:

  • The test provides results in 20 minutes and is easy to use
  • Connection to the OraSure Support Center for assistance and guidance

Convenient:

  • Pain free oral fluid-based test; no needles necessary
  • Can be used anytime, anywhere

Specifications

Content And Storage 2°C to27°C (36°F to 80°F)
CE Marker No
Clia Complexity Waived (OTC)
Detectable Analytes HIV-1, HIV-2
Sample Type Oral Fluid
Type HIV Test
Disposable Single-use
Test Time 20 min.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.