Learn More
OraSure Technologies OraQuick™ HIV Visual Reference Panel
SDP
Catalog No. 23222103 Shop All OraSure Technologies Products
Change view
Click to view available options
Quantity:
1 Set/Ea.
Catalog No. Quantity
23-222-103 1 Set/Ea.
1 options
This item is not returnable. View return policy
Catalog No. 23-222-103 Supplier OraSure Technologies Supplier No. 10010505
Add to Cart
Add to Cart
This item is not returnable. View return policy
The OraQuick™HIV Visual Reference Panel is a tool intended to assist operators in becoming proficient at reading specimens with antibody levels near the limit of detection of the device.
OraQuick™ HIV Visual Reference Panel
The foil pouch contains a package insert and three devices (one HIV Limit of Detection Device, one HIV Low Reactive Device and one HIV Non-Reactive Device) as described below:
  • HIV Limit of Detection Device
    • One OraQuick™ ADVANCE™ HIV-1/2 Rapid Antibody Test device that has been manufactured at a predetermined reactivity level to produce a reactive test result.
  • HIV Low Reactive Device
    • One OraQuick™ ADVANCE™ HIV-1/2 Rapid Antibody Test device that has been manufactured at a predetermined reactivity level to produce a reactive test result.
  • HIV Non-Reactive Device
    • One OraQuick™ ADVANCE™ HIV-1/2 Rapid Antibody Test device that has been manufactured to produce a non-reactive test result.
Specifications
Content And Storage 15°C to 30°C
Disposable Single-use
Quantity 1 Set/Ea.