Learn More
Med Vet International Oratene Drinking Water AdditIVe, 8oz Pump Bottle
Catalog No. 502828450
Encompass_Preferred
Contains Mutanase + Dextranase ensyme system-effectively inhibits and reduces plaque formation. Flavorless concentrate to add daily to drinking water-healthiest way to conveniently keep teeth clean without brushing. Does not contain chlorhexidine, chlorinesor alcohols. Removes plaque while inhibiting odor causing microbes. Convenient pump dosing system 100% safe for daily ingestion, cannot overdose, convenient to insure client compliance. Improves oral health. Two enzyme systems inhibit odor causing bacteria and reduces plaque biofilm formation. Comprehensive oral care system with antibacterial and antifungal properties. pH neutral.
Catalog No. 50-282-8450 Supplier Med Vet International Supplier No. PHVORATAD8
Contains Mutanase + Dextranase ensyme system-effectively inhibits and reduces plaque formation. Flavorless concentrate to add daily to drinking water-healthiest way to conveniently keep teeth clean without brushing. Does not contain chlorhexidine, chlorinesor alcohols. Removes plaque while inhibiting odor causing microbes. Convenient pump dosing system 100% safe for daily ingestion, cannot overdose, convenient to insure client compliance. Improves oral health. Two enzyme systems inhibit odor causing bacteria and reduces plaque biofilm formation. Comprehensive oral care system with antibacterial and antifungal properties. pH neutral.