Learn More
Bellco Glass, Inc. Orbital Shaker Complete, 115V-25x25
SDP
Catalog No. 501946130
Encompass_Preferred
The Bellco Benchtop Orbital Shakers are ideal for staining and washingelectrophoresis gels or for any applications requiring mixing or agitationof solutions. They are engineered to operate between the limits of 30 andapproximately 350 revolutions per minute, and to hold their speed at anyselected setting to approximately plus or minus five percent.. MADE IN THE USA--Speed range: 30 - 350 rpm +/- 5%--Circular orbit: 2.5 cm (1" ) orbit--Load: 11 kg (25 lbs) maximum--Weight: 7.6 kg (17 lbs)--7744-01010, 7744-10220 & 7744-01100 units have a platform deck of 25 x 25 cm--7744-02020, 7744-20220 & 7744-02200 units have a platform deck 50 x 50 cm
Catalog No. 50-194-6130 Supplier Bellco Glass, Inc. Supplier No. 774401010
The Bellco Benchtop Orbital Shakers are ideal for staining and washingelectrophoresis gels or for any applications requiring mixing or agitationof solutions. They are engineered to operate between the limits of 30 andapproximately 350 revolutions per minute, and to hold their speed at anyselected setting to approximately plus or minus five percent.. MADE IN THE USA--Speed range: 30 - 350 rpm +/- 5%--Circular orbit: 2.5 cm (1" ) orbit--Load: 11 kg (25 lbs) maximum--Weight: 7.6 kg (17 lbs)--7744-01010, 7744-10220 & 7744-01100 units have a platform deck of 25 x 25 cm--7744-02020, 7744-20220 & 7744-02200 units have a platform deck 50 x 50 cm