Learn More
Thomas Scientific Orbi-Blotter 14x12" platform, 115V
SDP
Catalog No. 501461267
Encompass
Gentle motion, for gel applications Large platform, 14x12” Incubator and cold room safe The Orbi-Blotter ™ is the a low speed orbital shaker designed for gel blotting, staining/destaining and washing applications. With accurate speed control down to as little as three rpm. The Orbi-Blotter features a gentle orbital motion with speed control down to as little as 3 rpm, making it the ideal orbital choice for low speed applications, such as blotting, washing, staining/destaining, etc. A large 14x12 in. platform is included along with a non-slip rubber mat for holding trays, dishes, plates and other flat vessels.
Catalog No. 50-146-1267 Supplier Thomas Scientific Supplier No. 1222U37
Gentle motion, for gel applications Large platform, 14x12” Incubator and cold room safe The Orbi-Blotter ™ is the a low speed orbital shaker designed for gel blotting, staining/destaining and washing applications. With accurate speed control down to as little as three rpm. The Orbi-Blotter features a gentle orbital motion with speed control down to as little as 3 rpm, making it the ideal orbital choice for low speed applications, such as blotting, washing, staining/destaining, etc. A large 14x12 in. platform is included along with a non-slip rubber mat for holding trays, dishes, plates and other flat vessels.