Learn More
Sigma Aldrich Fine Chemicals Biosciences Orbi Shaker JR. orbital sh1EA
Catalog No. 502908579
Encompass
The Benchmark Scientific Orbi-Shaker JR. a mini-sized replica molded after the larger Orbi-Shaker is a personal orbital shaker designed for smaller capacity applications and laboratories where space is at a premium. The shaker includes a standard 9.5 x 8 non-slip rubber flat platform that mixes at a 19mm orbit ideal for thoruoughly mixing and aerating tissue culture flasks Petri dishes staining trays and Erlenmeyer flasks.Additional platforms are also available for the Orbi-Shaker JR including the convenient MAGic Clamp platform for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.Mixing speed and run time are displayed on the large LED control panel located on the front side of the unit. The integral microprocessor includes a constant monitoring system that ensures accuracy through the duration of the program. Sophisticated over-temperature and over-speed control
Catalog No. 50-290-8579 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7424351EA
The Benchmark Scientific Orbi-Shaker JR. a mini-sized replica molded after the larger Orbi-Shaker is a personal orbital shaker designed for smaller capacity applications and laboratories where space is at a premium. The shaker includes a standard 9.5 x 8 non-slip rubber flat platform that mixes at a 19mm orbit ideal for thoruoughly mixing and aerating tissue culture flasks Petri dishes staining trays and Erlenmeyer flasks.Additional platforms are also available for the Orbi-Shaker JR including the convenient MAGic Clamp platform for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.Mixing speed and run time are displayed on the large LED control panel located on the front side of the unit. The integral microprocessor includes a constant monitoring system that ensures accuracy through the duration of the program. Sophisticated over-temperature and over-speed control